Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ATP6V1G3
Entrez Gene ID
424349
Gene Name
ATPase H+ transporting V1 subunit G3
Gene Aliases
-
Chromosome Location
Chr.8: 2500592 - 2511642 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_422192.4 XP_422192.1 2 - 3 338 68
GenBank mRNA BX932779.1 - 2 - 3 345 68
BX932960.1 - 1 - 2 105 68

View Full Product Details