Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GKN2
Entrez Gene ID
419515
Gene Name
gastrokine 2
Gene Aliases
GDDR
Chromosome Location
Chr.22: 406036 - 409430 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_417666.5 XP_417666.1 5 - 6 524 65
GenBank mRNA BX932799.1 - 5 - 6 488 65

View Full Product Details