Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NRSN1
Entrez Gene ID
420822
Gene Name
neurensin 1
Gene Aliases
VMP
Chromosome Location
Chr.2: 57938757 - 57945892 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_025147643.1 XP_025003411.1 2 - 3 242 92
XM_025147644.1 XP_025003412.1 2 - 3 1110 92
XM_418913.6 XP_418913.2 2 - 3 359 92
GenBank mRNA BX933919.1 - 2 - 3 381 92

View Full Product Details