Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NRSN1
Entrez Gene ID
420822
Gene Name
neurensin 1
Gene Aliases
VMP
Chromosome Location
Chr.2: 57938757 - 57945892 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BX933919.1 - 3 - 4 709 152

View Full Product Details