Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RPS24
Entrez Gene ID
423726
Gene Name
ribosomal protein S24
Gene Aliases
-
Chromosome Location
Chr.6: 14000232 - 14006196 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277343.1 NP_001264272.1 5 - 6 435 81
GenBank mRNA BX934192.1 - 4 - 5 409 81
BX934456.2 - 5 - 6 421 81

View Full Product Details