Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FRA10AC1
Entrez Gene ID
423803
Gene Name
FRA10A associated CGG repeat 1
Gene Aliases
C10orf4, F26C11.1-like, FRA10A
Chromosome Location
Chr.6: 21291693 - 21320994 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015288856.2 XP_015144342.1 3 - 4 292 70
GenBank mRNA BX934634.1 - 3 - 4 285 70

View Full Product Details