Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RIDA
Entrez Gene ID
426536
Gene Name
reactive intermediate imine deaminase A homolog
Gene Aliases
HRSP12, PSP
Chromosome Location
Chr.2: 127389771 - 127396467 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001234688.5 XP_001234689.4 4 - 5 320 118
GenBank mRNA BX932754.1 - 4 - 5 312 118
BX935048.1 - 4 - 5 294 118

View Full Product Details