Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TTC39C
Entrez Gene ID
421077
Gene Name
tetratricopeptide repeat domain 39C
Gene Aliases
C18orf17
Chromosome Location
Chr.2: 103247546 - 103281852 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015277984.2 XP_015133470.1 2 - 3 386 69
GenBank mRNA BX950535.1 - 2 - 3 282 69
CR405791.1 - 2 - 3 167 69

View Full Product Details