Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
INTS13
Entrez Gene ID
418213
Gene Name
integrator complex subunit 13
Gene Aliases
ASUN, C12orf11, C1H12ORF11
Chromosome Location
Chr.1: 67963167 - 67983180 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_416439.6 XP_416439.3 5 - 6 957 117
GenBank mRNA BX950662.1 - 5 - 6 635 117

View Full Product Details