Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMEM268
Entrez Gene ID
417248
Gene Name
transmembrane protein 268
Gene Aliases
C17H9ORF91, C9orf91
Chromosome Location
Chr.17: 2882256 - 2891735 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004945796.3 XP_004945853.2 1 - 2 141 152
XM_415524.6 XP_415524.4 1 - 2 149 152
GenBank mRNA CR353484.1 - 1 - 2 156 152

View Full Product Details