Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KNOP1
Entrez Gene ID
426994
Gene Name
lysine rich nucleolar protein 1
Gene Aliases
C14H16ORF88
Chromosome Location
Chr.14: 15091097 - 15101374 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004945463.3 XP_004945520.1 2 - 3 1515 154
XM_015294792.2 XP_015150278.1 2 - 3 1615 154
GenBank mRNA BX929426.2 - 1 - 2 1405 154

View Full Product Details