Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DLK1
Entrez Gene ID
423459
Gene Name
delta like non-canonical Notch ligand 1
Gene Aliases
-
Chromosome Location
Chr.5: 49117916 - 49144746 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142254.1 NP_001135726.1 3 - 4 285 92
XM_025150792.1 XP_025006560.1 7 - 8 1066 92
GenBank mRNA BX930935.2 - 3 - 4 276 92
EU288039.1 - 3 - 4 285 92

View Full Product Details