Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SKA2P1
Entrez Gene ID
417629
Gene Name
spindle and kinetochore associated complex subunit 2 pseudogene 1
Gene Aliases
SKA2
Chromosome Location
Chr.19: 7334925 - 7342348 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BX929961.1 - 1 - 2 421 144
BX933399.2 - 1 - 2 170 144

View Full Product Details