Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SYT12
Entrez Gene ID
422998
Gene Name
synaptotagmin 12
Gene Aliases
synaptotagmin-12
Chromosome Location
Chr.5: 1709714 - 1716080 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199512.1 NP_001186441.1 1 - 2 69 65
GenBank mRNA BX259616.3 - 1 - 2 70 65
BX936066.2 - 1 - 2 69 65

View Full Product Details