Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPCS2
Entrez Gene ID
419056
Gene Name
signal peptidase complex subunit 2
Gene Aliases
-
Chromosome Location
Chr.1: 196000701 - 196008838 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA CR386134.1 - 5 - 6 618 107

View Full Product Details