Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WRNIP1
Entrez Gene ID
395089
Gene Name
Werner helicase interacting protein 1
Gene Aliases
WHIP
Chromosome Location
Chr.2: 67588268 - 67613666 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_418979.6 XP_418979.2 5 - 6 1413 128
GenBank mRNA AB104724.1 - 4 - 5 708 128

View Full Product Details