Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SMAD1
Entrez Gene ID
395680
Gene Name
SMAD family member 1
Gene Aliases
-
Chromosome Location
Chr.4: 31056626 - 31187014 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001201455.1 NP_001188384.1 5 - 6 1253 85
XM_015276129.2 XP_015131615.2 6 - 7 1610 85
XM_015276130.2 XP_015131616.2 6 - 7 1637 85
XM_015276131.2 XP_015131617.2 6 - 7 1823 85
XM_015276132.2 XP_015131618.2 6 - 7 1503 85
XM_015276134.2 XP_015131620.1 5 - 6 2679 85
XM_025149512.1 XP_025005280.1 6 - 7 1784 85
XM_025149513.1 XP_025005281.1 11 - 12 2076 85
XM_025149514.1 XP_025005282.1 7 - 8 1718 85
GenBank mRNA AF143239.1 - 4 - 5 756 85
AY953143.1 - 5 - 6 1253 85
CV860526.1 - 4 - 5 634 85
DN854661.1 - 1 - 2 344 85

View Full Product Details