Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GLT8D2
Entrez Gene ID
418085
Gene Name
glycosyltransferase 8 domain containing 2
Gene Aliases
-
Chromosome Location
Chr.1: 54677179 - 54693103 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277383.2 NP_001264312.2 2 - 3 319 83
GenBank mRNA BU252180.1 - 2 - 3 319 83
BX929886.1 - 2 - 3 319 83

View Full Product Details