Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IFT22
Entrez Gene ID
417503
Gene Name
intraflagellar transport 22
Gene Aliases
RABL5
Chromosome Location
Chr.19: 3655950 - 3659541 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_415752.6 XP_415752.2 2 - 3 156 106
GenBank mRNA CR406254.1 - 2 - 3 140 106
CR406564.1 - 1 - 2 131 106

View Full Product Details