Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IQCC
Entrez Gene ID
419646
Gene Name
IQ motif containing C
Gene Aliases
-
Chromosome Location
Chr.23: 5310699 - 5313031 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BU382037.1 - 2 - 3 257 114
CR406781.1 - 2 - 3 258 114

View Full Product Details