Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MRPS18C
Entrez Gene ID
770813
Gene Name
mitochondrial ribosomal protein S18C
Gene Aliases
-
Chromosome Location
Chr.4: 46463751 - 46465919 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001252311.1 NP_001239240.1 1 - 2 250 104
GenBank mRNA BX932769.1 - 1 - 2 260 104

View Full Product Details