Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CARD19
Entrez Gene ID
415987
Gene Name
caspase recruitment domain family member 19
Gene Aliases
C12H9ORF89, C9orf89
Chromosome Location
Chr.12: 7033642 - 7054153 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001232398.5 XP_001232399.1 2 - 3 186 137
XM_001232443.5 XP_001232444.1 2 - 3 247 137
XM_004944551.3 XP_004944608.1 2 - 3 276 137
XM_004944552.3 XP_004944609.1 2 - 3 276 137
XM_015293413.2 XP_015148899.1 2 - 3 295 137
XM_015293414.2 XP_015148900.1 2 - 3 168 137
XM_015293415.2 XP_015148901.1 2 - 3 356 137
XM_414329.5 XP_414329.1 2 - 3 276 137
GenBank mRNA BX935395.1 - 2 - 3 203 137
CR338839.1 - 2 - 3 177 137

View Full Product Details