Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MURC
Entrez Gene ID
420986
Gene Name
muscle related coiled-coil protein
Gene Aliases
CAVIN4
Chromosome Location
Chr.2: 88737902 - 88751005 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_419073.5 XP_419073.2 1 - 2 878 103
GenBank mRNA BX935810.1 - 1 - 2 500 103

View Full Product Details