Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZFAND6
Entrez Gene ID
415483
Gene Name
zinc finger AN1-type containing 6
Gene Aliases
ZA20D3, ZGC:101121
Chromosome Location
Chr.10: 13022108 - 13071552 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277522.1 NP_001264451.1 3 - 4 456 81
NM_001277526.1 NP_001264455.1 3 - 4 397 81
NM_001277528.1 NP_001264457.1 2 - 3 248 81
XM_015291998.2 XP_015147484.1 3 - 4 366 81
XM_025153839.1 XP_025009607.1 3 - 4 283 81
XM_025153840.1 XP_025009608.1 2 - 3 191 81
GenBank mRNA BU109075.1 - 3 - 4 409 81
BU137192.1 - 2 - 3 198 81
BX935855.1 - 3 - 4 331 81
CF251301.1 - 2 - 3 250 81
CN237237.1 - 3 - 4 456 81
DR411810.1 - 3 - 4 395 81

View Full Product Details