Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPRED1
Entrez Gene ID
423292
Gene Name
sprouty related EVH1 domain containing 1
Gene Aliases
-
Chromosome Location
Chr.5: 30947874 - 31003437 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199709.1 NP_001186638.1 3 - 4 388 108
XM_015287344.2 XP_015142830.1 3 - 4 407 108
XM_015287345.2 XP_015142831.1 3 - 4 986 108
XM_015287346.2 XP_015142832.1 3 - 4 461 108
GenBank mRNA BX929595.2 - 2 - 3 348 108
BX950702.1 - 3 - 4 372 108

View Full Product Details