Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPRED1
Entrez Gene ID
423292
Gene Name
sprouty related EVH1 domain containing 1
Gene Aliases
-
Chromosome Location
Chr.5: 30947874 - 31003437 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199709.1 NP_001186638.1 4 - 5 445 102
XM_015287346.2 XP_015142832.1 4 - 5 518 102
GenBank mRNA BX929595.2 - 3 - 4 405 102
BX950702.1 - 4 - 5 429 102

View Full Product Details