Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GPN1
Entrez Gene ID
421980
Gene Name
GPN-loop GTPase 1
Gene Aliases
XAB1
Chromosome Location
Chr.3: 104286877 - 104302083 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_025149104.1 XP_025004872.1 1 - 2 114 85
XM_419990.3 XP_419990.2 1 - 2 423 85
GenBank mRNA CR352371.1 - 1 - 2 85 85

View Full Product Details