Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MRPS25
Entrez Gene ID
416129
Gene Name
mitochondrial ribosomal protein S25
Gene Aliases
-
Chromosome Location
Chr.12: 20125347 - 20137039 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277840.1 NP_001264769.1 1 - 2 220 138
GenBank mRNA BU349133.1 - 1 - 2 134 138
BX933213.2 - 1 - 2 181 138
CN231166.1 - 1 - 2 221 138

View Full Product Details