Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARPC3
Entrez Gene ID
416837
Gene Name
actin related protein 2/3 complex subunit 3
Gene Aliases
-
Chromosome Location
Chr.15: 5578016 - 5582602 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277486.1 NP_001264415.1 3 - 4 241 136
GenBank mRNA BX933901.2 - 3 - 4 310 136
BX934934.1 - 3 - 4 251 136
CN220727.1 - 3 - 4 241 136

View Full Product Details