Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AKIRIN2
Entrez Gene ID
421821
Gene Name
akirin 2
Gene Aliases
C6orf166, akirin-2
Chromosome Location
Chr.3: 76360041 - 76376531 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193595.2 NP_001180524.1 2 - 3 432 120
GenBank mRNA BX260396.3 - 2 - 3 457 120
BX934526.2 - 2 - 3 432 120
BX935737.2 - 3 - 4 294 120
CR387510.1 - 2 - 3 405 120
HM357352.1 - 2 - 3 342 120
KJ617061.1 - 2 - 3 342 120
KJ617062.1 - 2 - 3 342 120

View Full Product Details