The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_015286685.2 | XP_015142171.1 | 5 - 6 | 769 | 94 |
GenBank mRNA | BX934822.2 | - | 5 - 6 | 709 | 94 |