Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TSPAN6
Entrez Gene ID
422262
Gene Name
tetraspanin 6
Gene Aliases
TM4SF6
Chromosome Location
Chr.4: 5176802 - 5182610 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277603.1 NP_001264532.1 5 - 6 650 71
GenBank mRNA BX934878.2 - 5 - 6 544 71
BX935074.2 - 5 - 6 650 71

View Full Product Details