Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MYOM3
- Entrez Gene ID
-
419690
- Gene Name
- myomesin 3
- Gene Aliases
-
IFNLR1, IL28RA
- Chromosome Location
-
Chr.23: 5779176 - 5826654 on Build GRCg6a
- Gene Symbol
- IFNLR1
- Entrez Gene ID
-
419694
- Gene Name
- interferon lambda receptor 1
- Gene Aliases
-
CRF2-12, IL-28RA, IL28RA
- Chromosome Location
-
-
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
KF680104.1 |
- |
2 - 3 |
189 |
127 |
View Full Product Details