Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EGFL6
Entrez Gene ID
418637
Gene Name
EGF like domain multiple 6
Gene Aliases
-
Chromosome Location
Chr.1: 124845519 - 124907268 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277738.1 NP_001264667.1 - 295 89
GenBank mRNA BX930768.1 - - 229 89
BX950717.1 - - 295 89

View Full Product Details