Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HSD17B7
Entrez Gene ID
424367
Gene Name
hydroxysteroid 17-beta dehydrogenase 7
Gene Aliases
-
Chromosome Location
Chr.8: 4187219 - 4193089 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277506.1 NP_001264435.1 5 - 6 700 69
NM_001277507.1 NP_001264436.1 4 - 5 607 69
GenBank mRNA BU265534.1 - - 551 69
BX933486.2 - 5 - 6 654 69
BX935618.1 - 5 - 6 656 69
BX950661.1 - 4 - 5 593 69
CR352574.1 - 5 - 6 630 69

View Full Product Details