Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HSD17B7
Entrez Gene ID
424367
Gene Name
hydroxysteroid 17-beta dehydrogenase 7
Gene Aliases
-
Chromosome Location
Chr.8: 4187219 - 4193089 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277506.1 NP_001264435.1 6 - 7 802 80
NM_001277507.1 NP_001264436.1 5 - 6 709 80
GenBank mRNA BU265534.1 - - 653 80
BX933486.2 - 6 - 7 756 80
BX935618.1 - 6 - 7 758 80
BX950661.1 - 5 - 6 695 80
CR352574.1 - 6 - 7 732 80

View Full Product Details