Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MFSD12
Entrez Gene ID
420074
Gene Name
major facilitator superfamily domain containing 12
Gene Aliases
C19orf28, C28H19ORF28, PP3501
Chromosome Location
Chr.28: 1155052 - 1161689 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004948703.3 XP_004948760.1 5 - 6 928 86
XM_418193.6 XP_418193.3 5 - 6 909 86
GenBank mRNA CR352713.1 - 3 - 4 354 86

View Full Product Details