Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZNRF2
Entrez Gene ID
420678
Gene Name
zinc and ring finger 2
Gene Aliases
-
Chromosome Location
Chr.2: 41208589 - 41266458 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015281773.2 XP_015137259.1 4 - 5 1315 89
GenBank mRNA CR353109.1 - 4 - 5 415 89

View Full Product Details