Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ISM1
Entrez Gene ID
416740
Gene Name
isthmin 1
Gene Aliases
C20orf82, ISM, Isthmin, bA149I18.1, dJ1077I2.1, isthmin-1
Chromosome Location
Chr.3: 12824285 - 12880272 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015283524.2 XP_015139010.1 4 - 5 13190 94
XM_015283525.2 XP_015139011.1 4 - 5 1103 94
XM_415036.6 XP_415036.5 3 - 4 16735 94
GenBank mRNA CR353215.1 - 1 - 2 187 94

View Full Product Details