Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
METTL18
Entrez Gene ID
424400
Gene Name
methyltransferase like 18
Gene Aliases
MGC9084L
Chromosome Location
Chr.8: 5457117 - 5471915 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_422244.6 XP_422244.1 1 - 2 71
GenBank mRNA BX929354.2 - 1 - 2 29 71

View Full Product Details