Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RMDN1
Entrez Gene ID
420213
Gene Name
regulator of microtubule dynamics 1
Gene Aliases
FAM82B
Chromosome Location
Chr.2: 123036659 - 123049268 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277412.1 NP_001264341.1 7 - 8 770 90
GenBank mRNA BU465115.1 - 2 - 3 112 90
BX930716.1 - 7 - 8 722 90
BX932323.2 - 7 - 8 781 90

View Full Product Details