Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
N4BP2L1
Entrez Gene ID
418914
Gene Name
NEDD4 binding protein 2 like 1
Gene Aliases
-
Chromosome Location
Chr.1: 175887797 - 175900550 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_417110.6 XP_417110.2 4 - 5 711 60
GenBank mRNA BX933581.2 - 4 - 5 429 60

View Full Product Details