Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DPYS
Entrez Gene ID
420266
Gene Name
dihydropyrimidinase
Gene Aliases
-
Chromosome Location
Chr.2: 129839389 - 129867424 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277426.1 NP_001264355.1 6 - 7 1087 65
NM_001277427.1 NP_001264356.1 5 - 6 1015 65
XM_015282978.2 XP_015138464.1 5 - 6 911 65
GenBank mRNA BU381319.1 - 1 - 2 120 65
BX936207.2 - 5 - 6 1025 65
CO635756.1 - 6 - 7 1082 65

View Full Product Details