Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAJC12
Entrez Gene ID
423647
Gene Name
DnaJ heat shock protein family (Hsp40) member C12
Gene Aliases
-
Chromosome Location
Chr.6: 7337888 - 7349978 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199530.1 NP_001186459.1 4 - 5 615 86
XM_015288093.2 XP_015143579.1 5 - 6 1639 86
GenBank mRNA BX950610.2 - 5 - 6 540 86

View Full Product Details