Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BEND7
Entrez Gene ID
419037
Gene Name
BEN domain containing 7
Gene Aliases
C10orf30
Chromosome Location
Chr.1: 7324436 - 7372660 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282235.2 NP_001269164.1 5 - 6 1101 83
GenBank mRNA CR385344.1 - 1 - 2 70 83

View Full Product Details