Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PIEZO2
Entrez Gene ID
421052
Gene Name
piezo type mechanosensitive ion channel component 2
Gene Aliases
FAM38B
Chromosome Location
Chr.2: 97719562 - 98025242 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015282541.2 XP_015138027.2 54 - 55 8719 117
XM_015282543.2 XP_015138029.2 53 - 54 8551 117
XM_025147818.1 XP_025003586.1 54 - 55 8722 117
XM_025147819.1 XP_025003587.1 53 - 54 8610 117
XM_025147820.1 XP_025003588.1 54 - 55 8719 117
XM_025147821.1 XP_025003589.1 50 - 51 8085 117
XM_025147822.1 XP_025003590.1 53 - 54 8683 117
XM_025147824.1 XP_025003592.1 53 - 54 8554 117
GenBank mRNA CR385783.1 - 4 - 5 585 117

View Full Product Details