Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PMCH
Entrez Gene ID
418091
Gene Name
pro-melanin concentrating hormone
Gene Aliases
-
Chromosome Location
Chr.1: 55372262 - 55373651 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195795.1 NP_001182724.1 1 - 2 311 135
GenBank mRNA CR387227.1 - 1 - 2 173 135
HM853641.1 - 1 - 2 311 135

View Full Product Details