Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ISCA2
Entrez Gene ID
423352
Gene Name
iron-sulfur cluster assembly 2
Gene Aliases
HBLD1
Chromosome Location
Chr.5: 38118538 - 38136592 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BX930048.1 - 4 - 5 442 95

View Full Product Details