Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VAMP2
Entrez Gene ID
424827
Gene Name
vesicle-associated membrane protein 2 (synaptobrevin 2)
Gene Aliases
-
Chromosome Location
Chr.9: 5710328 - 5755795 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015277054.2 XP_015132540.1 4 - 5 364 80
XM_025153690.1 XP_025009458.1 4 - 5 637 80
GenBank mRNA BX930373.1 - 4 - 5 540 80

View Full Product Details