Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AMIGO2
Entrez Gene ID
417810
Gene Name
adhesion molecule with Ig like domain 2
Gene Aliases
-
Chromosome Location
Chr.1: 31370359 - 31379889 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199550.1 NP_001186479.1 2 - 2 1751 83
XM_015281376.2 XP_015136862.1 2 - 2 1878 83
GenBank mRNA BX932821.1 - 1 - 1 1149 83

View Full Product Details